missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-nitrophenol, 96%, Thermo Scientific™

Product Code. 11499893
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5460-31-1
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.137
MDL Number MFCD00007241
InChI Key GAKLFAZBKQGUBO-UHFFFAOYSA-N
Synonym 3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030
PubChem CID 79579
IUPAC Name 2-methyl-3-nitrophenol
SMILES CC1=C(C=CC=C1O)[N+](=O)[O-]

Specifications

Melting Point 148°C to 152°C
Odor Phenol-like
Quantity 1 g
UN Number UN2446
Beilstein 1946057
Formula Weight 153.14
Percent Purity 96%
Chemical Name or Material 2-Methyl-3-nitrophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'