missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(methylthio)pyrazine, 99%, Thermo Scientific™

Product Code. 11483964
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11483964 1 g 1g
11493964 5 g 5g
11403974 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11483964 Supplier Thermo Scientific Alfa Aesar Supplier No. A11957.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2882-20-4
Molecular Formula C6H8N2S
Molecular Weight (g/mol) 140.20
MDL Number MFCD00014610
InChI Key PPPFFGVGWFKTHX-UHFFFAOYSA-N
Synonym 2-methyl-3-methylthio pyrazine, 2-methylthio-3-methylpyrazine, 2-methyl-3-methylsulfanyl-pyrazine, pyrazine, 2-methyl-3-methylthio, 2-methylmercapto-3-methylpyrazine, methylthio methyl pyrazine, 3-methyl-2-methylthiopyrazine, 2-methyl-3-methylmercaptopyrazine, unii-ki350cs0lz, pyrazine, 2-methyl-3 or 5-methylthio
PubChem CID 76152
IUPAC Name 2-methyl-3-methylsulfanylpyrazine
SMILES CSC1=NC=CN=C1C

Specifications

Density 1.15
Boiling Point 213°C to 214°C
Flash Point 99°C (210°F)
Refractive Index 1.583
Quantity 1 g
Formula Weight 140.21
Percent Purity 99%
Chemical Name or Material 2-Methyl-3- (methylthio)pyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.