missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-3-furanethiol, ca.90%, balance oxidized compound, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 88.0 |
CAS Max % | 100.0 |
Color | Orange to Pink |
Density | 1.1450g/mL |
Boiling Point | 57°C to 60°C (44.0 mmHg) |
Flash Point | 37°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 88% min. (GC) |
Packaging | Glass bottle |
Refractive Index | 1.5100 to 1.55 |
Show More |
Safety and Handling
Product Identifier
- 2-Methyl-3-furanethiol, balance oxidized compound
- Danger
- Acute toxicity Category 2
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- H319-Causes serious eye irritation.
- H330-Fatal if inhaled.
- H301-Toxic if swallowed.
- H226-Flammable liquid and vapour.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'