missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methyl-3-buten-2-ol, 98%

Product Code. 11415696
Change view
Click to view available options
Quantity:
100mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11415696 100mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11415696 Supplier Alfa Aesar™ Supplier No. B20790.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 115-18-4
Boiling Point 98°C to 99°C
Flash Point 13°C (55°F)
Odor Like Alcohol
Molecular Formula C5H10O
MDL Number MFCD00004470
Quantity 100mL
UN Number UN1987
Beilstein 1698263
Synonym 2-methyl-3-buten-2-ol, 1,1-dimethylallyl alcohol, methylbutenol, 3-buten-2-ol, 2-methyl, 3-hydroxy-3-methyl-1-butene, dimethylvinylcarbinol, dimethylvinylmethanol, vinyldimethylcarbinol, isoprenyl alcohol, 3-methyl-1-buten-3-ol
InChI Key HNVRRHSXBLFLIG-UHFFFAOYSA-N
SMILES CC(C)(C=C)O
IUPAC Name 2-methylbut-3-en-2-ol
Molecular Weight (g/mol) 86.134
PubChem CID 8257
Formula Weight 86.13
Percent Purity 98%
Chemical Name or Material 2-Methyl-3-buten-2-ol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.