missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3,5-dinitrobenzoic acid, 97+%

Product Code. p-7021990
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 11491938

Brand: Thermo Scientific Alfa Aesar A14555.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 28169-46-2 | C8H6N2O6 | 226.14 g/mol

2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-3,5-dinitrobenzoic acid was used to synthesis heterocyclic derivatives and in the synthesis of isoquinolone.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28169-46-2
Molecular Formula C8H6N2O6
Molecular Weight (g/mol) 226.14
MDL Number MFCD00007161
InChI Key CDVNZMKTJIBBBV-UHFFFAOYSA-N
Synonym 2-methyl-3,5-dinitrobenzoic acid, 3,5-dinitro-2-methylbenzoic acid, 3,5-dinitro-o-toluic acid, benzoic acid, 2-methyl-3,5-dinitro, 3,5-dinitrotoluic acid, o-toluic acid, 3,5-dinitro, 2-methyl-3,5-dinitro-benzoic acid, 3,5-dinitro-2-methyl-benzoic acid, acmc-1cfeu, o-toluic acid,5-dinitro
PubChem CID 98824
IUPAC Name 2-methyl-3,5-dinitrobenzoic acid
SMILES CC1=C(C=C(C=C1[N+]([O-])=O)[N+]([O-])=O)C(O)=O

Specifications

Melting Point 203°C to 207°C
Quantity 25 g
Beilstein 2622906
Solubility Information Soluble in water.
Formula Weight 226.15
Percent Purity 98%
Chemical Name or Material 2-Methyl-3,5-dinitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.