missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-[(3,4-methylenedioxy)phenyl]propionic acid, Alfa Aesar™

Product Code. p-7030325
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11397529 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11397529 Supplier Thermo Scientific Alfa Aesar Supplier No. H27924.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
It is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool dry conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77269-66-0
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD06203229
InChI Key DBXAHUCZEXVUKS-UHFFFAOYSA-N
Synonym 2-methyl-3-3,4-methylenedioxy phenyl propionic acid, 3-2h-1,3-benzodioxol-5-yl-2-methylpropanoic acid, 3-1,3-benzodioxol-5-yl-2-methylpropanoic acid, 3-2h-benzo d 1,3-dioxolen-5-yl-2-methylpropanoic acid, acmc-20anr1, 1,3-benzodioxole-5-propanoicacid, a-methyl, alpha-methyl-1,3-benzodioxole-5-propionic acid, 3-1,3-benzodioxol-5-yl-2-methyl-propanoic acid, 1,3-benzodioxole-5-propanoic acid, .alpha.-methyl, alpha-methyl-1,?3-benzodioxole-?5-?propanoic acid
PubChem CID 19614615
IUPAC Name 3-(1,3-benzodioxol-5-yl)-2-methylpropanoic acid
SMILES CC(CC1=CC2=C(C=C1)OCO2)C(=O)O

Specifications

Melting Point 77°C to 80°C
Quantity 5 g
Solubility Information Slightly soluble in water.
Formula Weight 208.21
Chemical Name or Material 2-Methyl-3-[(3,4-methylenedioxy)phenyl]propionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.