missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-3-(2-furyl)propenal, 97%

Product Code. 11499372
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11499372 250 g 250g
11479372 10 g 10g
11489372 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11499372 Supplier Thermo Scientific Alfa Aesar Supplier No. A10125.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 874-66-8 | C8H8O2 | 136.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 874-66-8
Molecular Formula C8H8O2
Molecular Weight (g/mol) 136.15
MDL Number MFCD00063240
InChI Key ZNBXZUKDRRRQJK-FNORWQNLSA-N
Synonym 2-methyl-3-2-furyl propenal, alpha-methylfurylacrolein, 2-methyl-3-2-furyl acrolein, furfurylidene-2-propanal, furfurylidine-2-propanal, 2-methyl-3-furylacrolein, alpha-methyl-2-furanacrolein, 2-furfurylidenepropionaldehyde, 3-2-furyl methacrylaldehyde, 2-methyl-3 2-furyl acrolein
PubChem CID 6210220
IUPAC Name (E)-3-(furan-2-yl)-2-methylprop-2-enal
SMILES CC(=CC1=CC=CO1)C=O

Specifications

Density 1.097
Boiling Point 280°C to 281°C
Flash Point >100°C (212°F)
Refractive Index 1.614
Quantity 250 g
Beilstein 2438
Formula Weight 136.15
Percent Purity 97%
Chemical Name or Material 2-Methyl-3- (2-furyl)propenal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.