missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-vinyloxirane, 95%

Product Code. 11984761 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11984761 25 g 25g
11974761 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11984761 Supplier Thermo Scientific Acros Supplier No. 440680250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1838-94-4 | C5H8O | 84.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1838-94-4
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
InChI Key FVCDMHWSPLRYAB-UHFFFAOYSA-N
Synonym 2-methyl-2-vinyloxirane, isoprene monoxide, ccris 5685, 3,4-epoxy-3-methyl-1-butene, 2-methyl-2-vinyl-oxirane, oxirane, 2-ethenyl-2-methyl-, 2r, acmc-20m5m5, oxirane, 2-ethenyl-2-methyl, 4,4,4-3fmet-1-2-furyl-1,3-butandione
PubChem CID 92166
IUPAC Name 2-ethenyl-2-methyloxirane
SMILES CC1(CO1)C=C

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Colorless
Density 0.8000g/mL
Boiling Point 80.0°C to 81.0°C
Flash Point −8°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Quantity 25 g
Specific Gravity 0.8
Formula Weight 84.12
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-vinyloxirane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-vinyloxirane, 95%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.