missing translation for 'onlineSavingsMsg'
Learn More

2-(Methyl-2-pyridylamino)ethanol, 98%

Product Code. 11467236
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11467236 50 g 50g
11477236 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11467236 Supplier Thermo Scientific Alfa Aesar Supplier No. B25555.18

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 122321-04-4 | C8H12N2O | 152.197 g/mol

2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-pyridylaminoethanol is used as a reagent to synthesize Rosiglitazone (Maleate: R693500), a thiazolinedione drug used to reduce insulin resistance in patients with Type 2 diabetes.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 122321-04-4
Molecular Formula C8H12N2O
Molecular Weight (g/mol) 152.197
MDL Number MFCD02667633
InChI Key MWGKOPUDDQZERY-UHFFFAOYSA-N
Synonym 2-methyl pyridin-2-yl amino ethanol, 2-methyl-2-pyridylamino ethanol, 2-methyl pyridin-2-yl amino ethan-1-ol, 2-n-methyl-2-pyridylaminoethanol, 2-methyl-2-pyridinylamino ethanol, 2-n-methyl-2-pyridylamino ethanol, 2-n-methyl-n-2-pyridinyl amino ethanol, 2-n-2-hydroxyethyl-n-methylamino pyridine, 2-methyl 2-pyridinyl amino-1-ethanol
PubChem CID 737420
IUPAC Name 2-[methyl(pyridin-2-yl)amino]ethanol
SMILES CN(CCO)C1=CC=CC=N1

Specifications

Refractive Index 1.575
Quantity 50 g
Solubility Information Soluble in water.
Formula Weight 152.19
Percent Purity 98%
Chemical Name or Material 2-(Methyl-2-pyridylamino)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.