missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propen-1-ol, 98%

Product Code. 10153530 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
100 mL
500 mL
Unit Size:
100mL
10mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10153530 500 mL 500mL
10756361 10 mL 10mL
10773144 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10153530 Supplier Thermo Scientific Acros Supplier No. 150295000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 513-42-8 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-42-8
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004737
InChI Key BYDRTKVGBRTTIT-UHFFFAOYSA-N
Synonym methallyl alcohol, 2-methyl-2-propen-1-ol, 2-methylallyl alcohol, isopropenyl carbinol, 2-propen-1-ol, 2-methyl, beta-methallyl alcohol, methacryl alcohol, 3-hydroxy-2-methylpropene, beta-methylallyl alcohol, 2-methyl-2-propene-1-ol
PubChem CID 10557
IUPAC Name 2-methylprop-2-en-1-ol
SMILES CC(=C)CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -50°C
Density 0.8500g/mL
Boiling Point 113°C to 115°C
Flash Point 33°C
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula H2C=C(CH3)CH2OH
Refractive Index 1.4250 to 1.427
Quantity 500 mL
Beilstein 01, 443
Specific Gravity 0.85
Solubility Information Solubility in water: 11 %. Other solubilities: soluble in alcohols,esters,ketones and,hydrocarbons
Viscosity 2.24 mPa.s (20°C)
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-2-propen-1-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-propen-1-ol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.