missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-propanethiol, 99%

Product Code. 10205390 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 mL
500 mL
2.5 L
Packaging:
Glass bottle
Unit Size:
2.5 L
500mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10205390 2.5 L Glass bottle 2.5 L
10572731 50 mL Glass bottle 50mL
10467730 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10205390 Supplier Thermo Scientific Acros Supplier No. 149020025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 75-66-1 | C4H10S | 90.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-66-1
Molecular Formula C4H10S
Molecular Weight (g/mol) 90.18
MDL Number MFCD00004857
InChI Key WMXCDAVJEZZYLT-UHFFFAOYSA-N
Synonym 2-methyl-2-propanethiol, tert-butyl mercaptan, tert-butylthiol, tert-butanethiol, 2-propanethiol, 2-methyl, tert-butylmercaptan, t-butyl mercaptan, t-butylmercaptan, 2-isobutanethiol, tertiary-butyl mercaptan
PubChem CID 6387
IUPAC Name 2-methylpropane-2-thiol
SMILES CC(C)(C)S

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -1.1°C
Color Colorless
Density 0.8000g/mL
Boiling Point 62°C to 65°C
Flash Point -24°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)3CSH
Refractive Index 1.4220 to 1.424
Quantity 2.5 L
Beilstein 01, 383
Merck Index 15, 1582
Specific Gravity 0.8
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in liquid h2s,very soluble in alcohol and ether
Viscosity 0.638 mPa.s (20°C)
Formula Weight 90.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-propanethiol, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-propanethiol
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Flammable liquid Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H411-Toxic to aquatic life with long lasting effects.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.