missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-oxazoline, 99%

Product Code. 11470468
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11470468 100 g 100g
11460468 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11470468 Supplier Thermo Scientific Alfa Aesar Supplier No. B24612.22
  • ONLINE EXCLUSIVE PRICE
    £58.60
    £48.65 / 100g
    Save £9.95
    16% Off
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1120-64-5 | C4H7NO | 85.11 g/mol

2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N'-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N′-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

Solubility
Soluble in water (7051 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-64-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005298
InChI Key GUXJXWKCUUWCLX-UHFFFAOYSA-N
Synonym 2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
PubChem CID 70713
ChEBI CHEBI:53614
IUPAC Name 2-methyl-4,5-dihydro-1,3-oxazole
SMILES CC1=NCCO1

Specifications

Density 1.01
Boiling Point 109°C to 111°C
Flash Point 20°C (68°F)
Refractive Index 1.434
Quantity 100 g
UN Number UN1993
Beilstein 104227
Solubility Information Soluble in water (7051mg/L at 25°C).
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-oxazoline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.