missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-oxazoline, 99%

Code produit 11460468
Modifier l'affichage
Click to view available options
Quantity:
25 g
100 g
Conditionnement:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Code produit Quantity unitSize
11460468 25 g 25g
11470468 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Code produit 11460468 Fournisseur Thermo Scientific Alfa Aesar Code fournisseur B24612.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.

CAS: 1120-64-5 | C4H7NO | 85.11 g/mol

2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N'-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-oxazoline is used in cationic polymerization of a series of linear 2-alkyl-2-oxazolines under microwave irradiation. It undergoes polymerization with 2-butyl-2-oxazoline to form poly(2-oxazoline) block copolymer. It undergoes cationic ring-opening polymerization with 2-(dec-9-enyl)-2-oxazoline to yield copoly(2-oxazoline)s. It react with N′-phenyl-benzohydrazonoyl chloride to get 4-(2-hydroxyethyl)-5-methyl-1,3-diphenyl-1,2,4-triazol-4-ium chloride.

Solubility
Soluble in water (7051 mg/L at 25°C).

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 1120-64-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005298
InChI Key GUXJXWKCUUWCLX-UHFFFAOYSA-N
Synonym 2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
PubChem CID 70713
ChEBI CHEBI:53614
IUPAC Name 2-methyl-4,5-dihydro-1,3-oxazole
SMILES CC1=NCCO1

Spécification

Density 1.01
Boiling Point 109°C to 111°C
Flash Point 20°C (68°F)
Refractive Index 1.434
Quantity 25 g
UN Number UN1993
Beilstein 104227
Solubility Information Soluble in water (7051mg/L at 25°C).
Formula Weight 85.11
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-oxazoline
Afficher plus Afficher moins
compliance-icons
Identifiant produit
  • 2-Methyl-2-oxazoline
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable liquid Category 2
Mention de danger
  • H225-Highly flammable liquid and vapour.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Informations supplémentaires
  • MIXTURE LIST-Contain: 2-Methyl-2-oxazoline

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.