missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-nitropropane, 99%

Product Code. 10685632 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
25g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10685632 25g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10685632 Supplier Thermo Scientific Acros Supplier No. 182960250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 594-70-7 | C4H9NO2 | 103.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 594-70-7
CAS Min % 98.5
CAS Max % 100.0
Melting Point 24°C
Color White
Density 0.9500g/mL
Boiling Point 126°C to 127°C
Flash Point 19°C
Infrared Spectrum Authentic
Assay Percent Range 98.5 % min. (GC)
Packaging Glass bottle
Molecular Formula C4H9NO2
Linear Formula (CH3)3CNO2
Refractive Index 1.3990 to 1.403
MDL Number MFCD00007393
Quantity 25 g
Beilstein 01, 129
Specific Gravity 0.95
Synonym propane, 2-methyl-2-nitro, trimethylnitromethane, 2-nitroisobutane, 2-nitro-2-methylpropane, tert-nitrobutane, 1,1-dimethyl-1-nitroethane, nitro-tert-butane, 2-methyl-2-nitro-propane, unii-9dkh5l679z, ccris 5044
Solubility Information Solubility in water: insoluble
InChI Key AIMREYQYBFBEGQ-UHFFFAOYSA-N
SMILES CC(C)(C)[N+](=O)[O-]
IUPAC Name 2-methyl-2-nitropropane
Molecular Weight (g/mol) 103.12
PubChem CID 11672
Formula Weight 103.12
Percent Purity 99%
Physical Form Crystalline Powder, Crystals or Flakes
Chemical Name or Material 2-Methyl-2-nitropropane, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-nitropropane
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.