missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-imidazoline, 95%, Thermo Scientific Chemicals

Catalog Number 11966271
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
534-26-9
C4H8N2
84.12
VWSLLSXLURJCDF-UHFFFAOYSA-N
2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
10798
2-methyl-4,5-dihydro-1H-imidazole
CC1=NCCN1
This item is not returnable. View return policy
534-26-9
C4H8N2
84.12
VWSLLSXLURJCDF-UHFFFAOYSA-N
2-methyl-2-imidazoline, 2-methylimidazoline, 1h-imidazole, 4,5-dihydro-2-methyl, methylglyoxalidine, 2-imidazoline, 2-methyl, 4,5-dihydro-2-methyl-1h-imidazole, unii-987f50e3pe, 2-methyl imidazoline, 2-methyl-4,5-dihydroimidazole, 2-methyl-20imidazolin
10798
2-methyl-4,5-dihydro-1H-imidazole
CC1=NCCN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Melting Point 104°C to 107°C
Color Colorless to Yellow or White to Yellow
Boiling Point 110°C (2.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 5 g
Merck Index 15,5696
Formula Weight 84.12
Percent Purity 95%
Physical Form Crystals, Powder or Needles
Chemical Name or Material 2-Methyl-2-imidazoline
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2-imidazoline
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'