missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-2-butanol, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Description
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.
Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.
Notes
Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Melting Point | -12°C |
Density | 0.805 g/mL |
Boiling Point | 101°C to 103°C |
Flash Point | 20°C (68°F) |
Assay Percent Range | 0.98 |
Refractive Index | 1.405 |
Quantity | 500 mL |
UN Number | UN1105 |
Beilstein | 1361351 |
Merck Index | 14,7140 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'