missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butanol, 98%

Product Code. p-3172645
Change view
Click to view available options
Quantity:
10,000 mL
2500 mL
500 mL
Unit Size:
10 L
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433857 500 mL 500mL
11443857 2500 mL 2500mL
11339589 10,000 mL 10 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11433857 Supplier Thermo Scientific Alfa Aesar Supplier No. A18304.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 75-85-4 | C5H12O | 88.15 g/mol

2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-2-butanol is used as a solvent in flavors, pharmaceuticals, corrosion inhibitors, resins, gums, coating materials and in organic synthesis. It acts as a frothing agent (ore flotation), surfactant (petroleum recovery) and stabilizing agent. Further, it is used in processing aids.

Solubility
Miscible with alcohol, ether, benzene, chloroform, glycerol, oils and acetone.

Notes
Light sensitive. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-85-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004478
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
PubChem CID 6405
IUPAC Name 2-methylbutan-2-ol
SMILES CCC(C)(C)O

Specifications

Melting Point -12°C
Density 0.805 g/mL
Boiling Point 101°C to 103°C
Flash Point 20°C (68°F)
Assay Percent Range 0.98
Refractive Index 1.405
Quantity 500 mL
UN Number UN1105
Beilstein 1361351
Merck Index 14,7140
Solubility Information Miscible with alcohol,ether,benzene,chloroform,glycerol,oils and acetone.
Formula Weight 88.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-butanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methyl-2-butanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.