missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2,4-pentanediol, 99%

Product Code. 10772314 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum
Unit Size:
1 L
2.5 L
25 L
This item is not returnable. View return policy

Product Code. 10772314

Brand: Thermo Scientific Acros 150340025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 107-41-5 | C6H14O2 | 118.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-41-5
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.18
MDL Number MFCD00004547
InChI Key SVTBMSDMJJWYQN-UHFFFAOYSA-N
Synonym hexylene glycol, 2-methyl-2,4-pentanediol, pinakon, 2,4-dihydroxy-2-methylpentane, 2,4-pentanediol, 2-methyl, diolane, isol, 4-methyl-2,4-pentanediol, 1,1,3-trimethyltrimethylenediol, 2-methyl pentane-2,4-diol
PubChem CID 7870
ChEBI CHEBI:62995
IUPAC Name 2-methylpentane-2,4-diol
SMILES CC(CC(C)(C)O)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -40.0°C
Density 0.9220g/mL
Boiling Point 197.0°C
Flash Point 93°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass Bottle
Linear Formula CH3CH(OH)CH2C(CH3)2OH
Refractive Index 1.4260 to 1.4280
Quantity 2.5 L
Beilstein 01, 486
Fieser 01,693
Merck Index 15, 4746
Specific Gravity 0.922
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,ether and lower aliphathic,hydrocarbons
Viscosity 36 mPa.s (20°C)
Formula Weight 118.18
Percent Purity 99%
Physical Form Slightly Viscous Liquid
Chemical Name or Material 2-Methyl-2, 4-pentanediol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-2,4-pentanediol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2-methyl-2,4-Pentanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.