missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-imidazole-4-carbothioamide, 97%, Thermo Scientific™

Product Code. 10652073
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10652073 1 g 1g
10009471 250 mg 250mg
11360747 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10652073 Supplier Thermo Scientific Maybridge Supplier No. SB02117DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 129486-91-5
Molecular Formula C5H7N3S
Molecular Weight (g/mol) 141.192
MDL Number MFCD03659724
InChI Key MNYPQSNAWPZXRV-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbothioamide, 1h-imidazole-5-carbothioamide,2-methyl, 1h-imidazole-5-carbothioamide, 2-methyl, acmc-1c27g, amino 2-methylimidazol-4-yl methane-1-thione, 2-methyl-3h-imidazole-4-carbothioamide
PubChem CID 2796714
IUPAC Name 2-methyl-1H-imidazole-5-carbothioamide
SMILES CC1=NC=C(N1)C(=S)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 141.2
Percent Purity 97%
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbothioamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.