missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-imidazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10192934
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10192934 10 g Amber Glass Bottle 10g
10693413 1 g Amber Glass Bottle 1g
10549434 25 g Amber Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10192934 Supplier Thermo Scientific Maybridge Supplier No. SB01361EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 35034-22-1
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
InChI Key ZWULFIBGPXWGFG-UHFFFAOYSA-N
Synonym 2-methyl-1h-imidazole-4-carbaldehyde, 2-methyl-4-formylimidazole, 2-methylimidazole-4-carbaldehyde, 4-formyl-2-methyl-1h-imidazole, 2-methylimidazole-5-carboxaldehyde, 2-methyl-3h-imidazole-4-carbaldehyde, 2-methyl-1h-imidazole-4-carboxaldehyde, 4-formyl-2-methylimidazole, 2-methyl-4-imidazolecarboxaldehyde, 2-methyl-1h-imidazole-5-carboxaldehyde
PubChem CID 2795878
IUPAC Name 2-methyl-1H-imidazole-5-carbaldehyde
SMILES CC1=NC=C(N1)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 110.12
Chemical Name or Material 2-Methyl-1H-imidazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.