missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester, 97%

Product Code. 11936751 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11936751 1 g 1g
11946751 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11936751 Supplier Thermo Scientific Acros Supplier No. 445160010
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1314216-34-6 | C14H19BN2O2 | 258.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314216-34-6
Molecular Formula C14H19BN2O2
Molecular Weight (g/mol) 258.13
MDL Number MFCD13176536
InChI Key CPQGLMCLTALRHF-UHFFFAOYSA-N
Synonym 2-methyl-1h-benzimidazole-5-boronic acid pinacol ester, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzo d imidazole, cpqglmcltalrhf-uhfffaoysa-n, 2-methyl-1h-benzimidazole-5-boronic acidpinacol ester, 2-methyl-1h-benzimidazole-5-boronic acid, pinacol ester, 1h-benzimidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 2-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-1h-benzo d imidazole, 2-methyl-6-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-benzimidazole
PubChem CID 46738035
IUPAC Name 2-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-1,3-benzodiazole
SMILES CC1=NC2=CC(=CC=C2N1)B1OC(C)(C)C(C)(C)O1

Specifications

Melting Point 211°C to 212°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 258.13
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 2-Methyl-1H-benzimidazole-5-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.