missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-1-tetralone, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
10 g
Description
Methylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Methylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Methylation of 1-tetralone followed by the subsequent dehydrogenation of 2-methyl-1-tetralone in the presence of Pd/C catalyst also has been reported.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Density | 1.058 |
Boiling Point | 129°C (12 mmHg) |
Flash Point | >110°C (230°F) |
Refractive Index | 1.552 |
Quantity | 1 g |
Solubility Information | Insoluble in water. |
Formula Weight | 160.22 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methyl-1-tetralone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'