missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pyrroline, Thermo Scientific Chemicals

Product Code. 11376259
Change view
Click to view available options
Quantity:
5 mL
25 mL
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376259 5 mL 5mL
11386259 25 mL 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11376259 Supplier Thermo Scientific Alfa Aesar Supplier No. 044853.AA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

2-Methyl-1-pyrroline acts as monocyclic imine. It is a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-32-2
Molecular Formula C5H10N
Molecular Weight (g/mol) 84.14
MDL Number MFCD00128806
InChI Key OZLJZWIVBYLSOK-UHFFFAOYSA-N
Synonym 2-methyl-1-pyrroline, 3,4-dihydro-5-methyl-2h-pyrrole, 1pyrroline2methyl, 2h-pyrrole, 3,4-dihydro-5-methyl, pubchem9412
PubChem CID 70103
ChEBI CHEBI:78856
IUPAC Name 5-methyl-3,4-dihydro-2H-pyrrole
SMILES C[C+]1CCCN1

Specifications

Density 0.878g/mL
Boiling Point 104°C to 105°C
Flash Point 10°C (50°F)
Quantity 5 mL
UN Number UN2733
Formula Weight 83.13
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-pyrroline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.