missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol (Isobutyl Alcohol), TRC

Product Code. 30133186
Change view
Click to view available options
Quantity:
50 mL
250 mL
Unit Size:
250mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133186 250 mL 250mL
17985105 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133186 Supplier Toronto Research Chemicals Supplier No. TRCI780075250ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4 H10 O
Molecular Weight (g/mol) 74.12
Synonym 1-Propanol, 2-methyl-, Isobutyl alcohol (8CI), 2-Methyl-1-propanol, 1-Hydroxy-2-methylpropane, 1-Isobutanol, 1-Methyl-2-propanol, 2-Methylpropyl alcohol, Isobutanol, Isopropyl carbinol, NSC 5708, iso-Butyl alcohol, 2-Methylpropanol
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Quantity 250 mL
InChI Formula InChI=1S/C4H10O/c1-4(2)3-5/h4-5H,3H2,1-2H3
Formula Weight 74.0732
Chemical Name or Material Isobutanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.