missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-propanol, 99+%, spectrophotometric grade

Product Code. 10367920 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10367920 2.5 L Glass bottle 2.5 L
10514111 500 mL Glass bottle 500mL
10357920 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10367920 Supplier Thermo Scientific Acros Supplier No. 167700025
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 78-83-1 | C4H10O | 74.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.12
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Absorbance 0.01 max. at 330nm, 0.1 max. at 250nm, 0.5 max. at 230nm, 1 max. at 220nm
CAS Min % 99.0
CAS Max % 100.0
Melting Point -108.0°C
Density 0.8000g/mL
Boiling Point 108.0°C
Flash Point 28°C
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula (CH3)2CHCH2OH
Refractive Index 1.3945 to 1.3965
Quantity 2.5 L
Beilstein 01, 373
Residue after Evaporation 0.0005% max.
Merck Index 15, 5176
Specific Gravity 0.8
Solubility Information Solubility in water: 95g/L (20°C). Other solubilities: soluble in alcohol, ether, acetone
Viscosity 4 Pas (20°C)
Formula Weight 74.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1-propanol, 99+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-1-propanol
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.