missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-Propanol, ≥99.0%, ACS Reagent, Honeywell Riedel-de Haën™

Product Code. p-7120069
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 15623630

Brand: Honeywell-Riedel-de Haen 320048500ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.0%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-83-1
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00004740
InChI Key ZXEKIIBDNHEJCQ-UHFFFAOYSA-N
Synonym 2-methyl-1-propanol, isobutanol, isobutyl alcohol, 1-propanol, 2-methyl, 1-hydroxymethylpropane, isopropylcarbinol, iso-butyl alcohol, 2-methylpropyl alcohol, isobutylalkohol, i-butyl alcohol
PubChem CID 6560
ChEBI CHEBI:46645
IUPAC Name 2-methylpropan-1-ol
SMILES CC(C)CO

Specifications

Melting Point -108°C
Density 0.803g/mL at 25°C
Boiling Point 108°C
Flash Point 28°C (82.4°F)
Ignition Point 801°F
Packaging Glass Bottle
Linear Formula (CH3)2CHCH2OH
Quantity 500 mL
UN Number UN1212
Beilstein 1730878
Vapor Density 2.55 (vs air)
Formula Weight 74.12g/mol
Vapor Pressure 8mmHg (20°C), 8.8mmHg (0°C)
Percent Purity ≥99.0%
Grade ACS Reagent
Chemical Name or Material 2-Methyl-1-Propanol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.