missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-piperidino-2-propanamine, 97%, Thermo Scientific™

Product Code. 10689344
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 6105-74-4
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.27
MDL Number MFCD05022423
InChI Key IMBNEKOMEDCTCL-UHFFFAOYSA-N
Synonym 2-methyl-1-piperidin-1-yl propan-2-amine, 2-methyl-1-piperidino-2-propanamine, 1,1-dimethyl-2-piperidin-1-yl-ethylamine, 1,1-dimethyl-2-piperidin-1-ylethyl amine, 2-methyl-1-piperidin-1-yl-2-propanamine, 1,1-dimethyl-2-piperidinoethylamin, 1-piperidineethanamine,a,a-dimethyl, 1,1-dimethyl-2-piperidin-1-ylethylamine
PubChem CID 2794702
IUPAC Name 2-methyl-1-piperidin-1-ylpropan-2-amine
SMILES CC(C)(N)CN1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-piperidino-2-propanamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'