missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-piperidino-2-propanamine, 97%, Thermo Scientific™

Product Code. 10421673
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10421673 1 g 1g
10689344 250 mg 250mg
10163324 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10421673 Supplier Thermo Scientific Maybridge Supplier No. MO01185DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 6105-74-4
Molecular Formula C9H20N2
Molecular Weight (g/mol) 156.27
MDL Number MFCD05022423
InChI Key IMBNEKOMEDCTCL-UHFFFAOYSA-N
Synonym 2-methyl-1-piperidin-1-yl propan-2-amine, 2-methyl-1-piperidino-2-propanamine, 1,1-dimethyl-2-piperidin-1-yl-ethylamine, 1,1-dimethyl-2-piperidin-1-ylethyl amine, 2-methyl-1-piperidin-1-yl-2-propanamine, 1,1-dimethyl-2-piperidinoethylamin, 1-piperidineethanamine,a,a-dimethyl, 1,1-dimethyl-2-piperidin-1-ylethylamine
PubChem CID 2794702
IUPAC Name 2-methyl-1-piperidin-1-ylpropan-2-amine
SMILES CC(C)(N)CN1CCCCC1

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 156.27
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-piperidino-2-propanamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.