missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-1-propanol, 98%

Product Code. 11485586
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11485586

Brand: Thermo Scientific Alfa Aesar B20729.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-69-8 | C10H14O | 150.221 g/mol

It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker's yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the large-scale preparation of (R)-and (S)-1-(2, 4, 6-triisopropylphenyl) ethanol and also in the baker′s yeast mediated asymmetric reduction of cinnamaldehyde derivatives.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-69-8
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00065000
InChI Key GMDYDZMQHRTHJA-UHFFFAOYSA-N
Synonym 2-methyl-1-phenyl-1-propanol, .alpha.-isopropylbenzyl alcohol, isopropyl benzyl alcohol, 2-methyl-1-phenylpropanol, benzenemethanol, .alpha.-1-methylethyl, 1-phenyl-2-methylpropyl alcohol, 1r-2-methyl-1-phenyl-propan-1-ol, phenylisopropylcarbinol, isopropylphenylcarbinol, acmc-20anr9
PubChem CID 95626
IUPAC Name 2-methyl-1-phenylpropan-1-ol
SMILES CC(C)C(C1=CC=CC=C1)O

Specifications

Density 0.964
Boiling Point 124°C to 125°C (15 mmHg)
Flash Point 86°C (187°F)
Refractive Index 1.513
Quantity 5 g
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 150.22
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-phenyl-1-propanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.