missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-pentene, 97%

Product Code. 15400128
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Unit Size:
100mL
25mL
5mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15400128 5 mL 5mL
15410128 25 mL 25mL
15420128 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15400128 Supplier Thermo Scientific Alfa Aesar Supplier No. H53373.AA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 763-29-1 | C6H12 | 84.162 g/mol

2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-pentene is prepared from 2-Methyl-1-pentene. 2-methyl-1-pentene is used in copolymerization of ethylene catalyzed by nonbridged half-titanocenes. The isomerization of 2-methyl-1-pentene leads to anionic activation of c-h bonds in olefins.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-29-1
Molecular Formula C6H12
Molecular Weight (g/mol) 84.162
MDL Number MFCD00009405
InChI Key WWUVJRULCWHUSA-UHFFFAOYSA-N
Synonym 2-methyl-1-pentene, 2-methylpentene, 1-pentene, 2-methyl, 4-methyl-4-pentene, 2-methyl-pentene-1, unii-okc25o38mc, okc25o38mc, acmc-1bixb, 4-01-00-00841 beilstein handbook reference, c2h5ch2c ch3 =ch2
PubChem CID 12986
IUPAC Name 2-methylpent-1-ene
SMILES CCCC(=C)C

Specifications

Melting Point -136°C
Density 0.682
Boiling Point 62°C
Flash Point −27°C (−17°F)
Refractive Index 1.392
Quantity 5 mL
UN Number UN2288
Beilstein 1340499
Solubility Information Insoluble in water.
Formula Weight 84.16
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-pentene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.