missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-2-Methyl-1-butanol, 98%

Product Code. 11494857
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494857 500 mL 500mL
11349589 100 mL 100mL
11404867 2500 mL 2500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11494857 Supplier Thermo Scientific Alfa Aesar Supplier No. B21825.AP
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-32-6 | C5H12O | 88.15 g/mol

2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-butanol is a volatile metabolite produced by a number of different plant species. Used as a solvent in organic synthesis (introduction of active amyl group), in lubricants, plasticizers, additives for oils & paints. It is also employed as a perfuming agent.

Solubility
Soluble in water. [183 g/L (0 C)] and alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 137-32-6
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004743
InChI Key QPRQEDXDYOZYLA-UHFFFAOYSA-N
Synonym 2-methyl-1-butanol, active amyl alcohol, 1-butanol, 2-methyl, 2-methylbutanol, sec-butylcarbinol, dl-2-methyl-1-butanol, 2-methyl-n-butanol, 2-methylbutyl alcohol, 2-methyl butanol-1, primary active amyl alcohol
PubChem CID 8723
ChEBI CHEBI:48945
IUPAC Name 2-methylbutan-1-ol
SMILES CCC(C)CO

Especificaciones

Melting Point <-70°C
Density 0.815
Boiling Point 129°C to 131°C
Flash Point 43°C (109°F)
Refractive Index 1.411
Quantity 500 mL
UN Number UN1105
Beilstein 1718810
Merck Index 14,6030
Solubility Information Soluble in water. [183g/L (0 C)] and alcohol.
Formula Weight 88.15
Percent Purity 98%
Chemical Name or Material (+/-)-2-Methyl-1-butanol
Mostrar más Mostrar menos
compliance-icons compliance-icons
Nombre del producto
  • (+/-)-2-Methyl-1-butanol
Palabra de advertencia
  • Warning
Clasificación
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Indicaciones de peligro
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Consejos de prudencia
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Otros peligros
  • MIXTURE LIST-Contain: (+/-)-2-Methyl-1-butanol

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.