missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10187814
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10187814 5 g 5g
10009561 1 g 1g
10691693 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10187814 Supplier Thermo Scientific Maybridge Supplier No. SP00294DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 109812-64-8
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
MDL Number MFCD01567220
InChI Key DWIYTBRYOQDHTE-UHFFFAOYSA-N
Synonym 2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylic acid, 1,5-diphenyl-2-methylpyrrole-3-carboxylic acid, 2-methyl-1,5-diphenyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl, maybridge1_008589, acmc-20mclm, 2-methyl-1,5-diphenyl-pyrrole-3-carboxylic acid
PubChem CID 2735512
IUPAC Name 2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
SMILES CC1=C(C=C(N1C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 5 g
Formula Weight 277.32
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.