missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,5-diphenyl-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10691693
Click to view available options
Quantity:
1 g
5 g
10 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 109812-64-8
Molecular Formula C18H15NO2
Molecular Weight (g/mol) 277.323
MDL Number MFCD01567220
InChI Key DWIYTBRYOQDHTE-UHFFFAOYSA-N
Synonym 2-methyl-1,5-diphenyl-1h-pyrrole-3-carboxylic acid, 1,5-diphenyl-2-methylpyrrole-3-carboxylic acid, 2-methyl-1,5-diphenyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1,5-diphenyl, maybridge1_008589, acmc-20mclm, 2-methyl-1,5-diphenyl-pyrrole-3-carboxylic acid
PubChem CID 2735512
IUPAC Name 2-methyl-1,5-diphenylpyrrole-3-carboxylic acid
SMILES CC1=C(C=C(N1C2=CC=CC=C2)C3=CC=CC=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 10 g
Formula Weight 277.32
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 5-diphenyl-1H-pyrrole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'