missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-thiazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10021214
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10021214 5 g 5g
10471684 250 mg 250mg
10644385 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021214 Supplier Thermo Scientific Maybridge Supplier No. CC05704DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20949-84-2
Molecular Formula C5H5NOS
Molecular Weight (g/mol) 127.16
MDL Number MFCD06655197
InChI Key AEHWVNPVEUVPMT-UHFFFAOYSA-N
PubChem CID 2763191
IUPAC Name 2-methyl-1,3-thiazole-4-carbaldehyde
SMILES CC1=NC(C=O)=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 5 g
Synonym 4-formyl-2-methylthiazole, 2-methylthiazole-4-carbaldehyde, 2-methylthiazole-4-carboxaldehyde, 4-thiazolecarboxaldehyde, 2-methyl, 2-methyl-4-thiazolecarboxaldehyde, 2-methyl-4-formylthiazole, 2-methyl-4-fomylthiazole, 4-formyl-2-methyl-1,3-thiazole, 4-thiazolecarboxaldehyde,2-methyl, 2-methyl-1,3-thiazole-4-carboxaldehyde
Formula Weight 127.17
Percent Purity 97%
Chemical Name or Material 2-Methyl-1, 3-thiazole-4-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.