missing translation for 'onlineSavingsMsg'
Learn More

(2-Methyl-1,3-thiazol-4-yl)methylamine, 97%, Thermo Scientific™

Product Code. 10208513
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10208513 1 g Amber Glass Bottle 1g
10613595 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10208513 Supplier Thermo Scientific Maybridge Supplier No. CC05713DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 103694-26-4
Molecular Formula C5H8N2S
Molecular Weight (g/mol) 128.193
MDL Number MFCD06212804
InChI Key ZCKAEFOHSOQKHN-UHFFFAOYSA-N
Synonym 2-methylthiazol-4-yl methanamine, 2-methyl-1,3-thiazole-4-methylamine, 2-methyl-1,3-thiazol-4-yl methanamine, 2-methyl-1,3-thiazol-4-yl methylamine, c-2-methyl-thiazol-4-yl-methylamine, 4-thiazolemethanamine, 2-methyl, 4-thiazolemethanamine,2-methyl, 4-aminomethyl-2-methyl-1,3-thiazole, 1-2-methyl-1,3-thiazol-4-yl methanamine, 2-methylthiazol-4-yl methylamine
PubChem CID 18467479
IUPAC Name (2-methyl-1,3-thiazol-4-yl)methanamine
SMILES CC1=NC(=CS1)CN

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 128.2
Percent Purity 97%
Chemical Name or Material (2-Methyl-1, 3-thiazol-4-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.