missing translation for 'onlineSavingsMsg'
Learn More

2-methyl-1,3-oxazole-4-carbaldehyde, Thermo Scientific™

Catalog Number 11356975
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
113732-84-6
C5H5NO2
111.1
ARAUEWKXKTYCHZ-UHFFFAOYSA-N
2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde
11073372
2-methyl-1,3-oxazole-4-carbaldehyde
CC1=NC(=CO1)C=O
This item is not returnable. View return policy
113732-84-6
C5H5NO2
111.1
ARAUEWKXKTYCHZ-UHFFFAOYSA-N
2-methyloxazole-4-carbaldehyde, 2-methyl-oxazole-4-carbaldehyde, 2-methyloxazole-4-carboxaldehyde, 2-methyl-4-formyloxazole, 4-oxazolecarboxaldehyde, 2-methyl, 4-formyl-2-methyl-1,3-oxazole, 2-methyl-1,3-oxazole-4-carboxaldehyde, acmc-20ai0d, 2-methyl-4-oxazolecarbaldehyde
11073372
2-methyl-1,3-oxazole-4-carbaldehyde
CC1=NC(=CO1)C=O
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 111.1
Percent Purity 97%
Chemical Name or Material 2-methyl-1, 3-oxazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'