missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-(3-morpholinopropyl)-5-phenyl-1H-pyrrole-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10283522
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10283522 1 g Amber Glass Bottle 1g
10355302 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10283522 Supplier Thermo Scientific Maybridge Supplier No. SP00344DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306936-20-9
Molecular Formula C19H24N2O3
Molecular Weight (g/mol) 328.412
MDL Number MFCD01567236
InChI Key OZRPKALPSGYEHX-UHFFFAOYSA-N
Synonym 2-methyl-1-3-morpholinopropyl-5-phenyl-1h-pyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylicacid, 2-methyl-1-3-4-morpholinyl propyl-5-phenyl, 2-methyl-1-3-morpholin-4-yl propyl-5-phenyl-1h-pyrrole-3-carboxylic acid, 2-methyl-1-3-morpholin-4-yl propyl-5-phenylpyrrole-3-carboxylic acid, 2-methyl-1-3-morpholin-4-yl prop-1-yl-5-phenyl-1h-pyrrole-3-carboxylic acid, maybridge1_008371, 2-methyl-1-3-morpholin-4-ylpropyl-5-phenyl-1h-pyrrole-3-carboxylic acid, 2-methyl-1-3-morpholin-4-ylpropyl-5-phenylpyrrole-3-carboxylic acid
PubChem CID 2736904
IUPAC Name 2-methyl-1-(3-morpholin-4-ylpropyl)-5-phenylpyrrole-3-carboxylic acid
SMILES CC1=C(C=C(N1CCCN2CCOCC2)C3=CC=CC=C3)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 328.41
Percent Purity 97%
Chemical Name or Material 2-Methyl-1-(3-morpholinopropyl)-5-phenyl-1H-pyrrole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.