missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dithiane, 98%

Product Code. 11372705
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372705 5 g 5g
11362705 1 g 1g
11382705 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11372705 Supplier Thermo Scientific Alfa Aesar Supplier No. L00209.06
Add to basket
This item is not returnable. View return policy

CAS: 6007-26-7 | C5H10S2 | 134.255 g/mol

It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It has been employed in the synthesis of methyl ketones. 2-Methyl-1,3-dithiane is suitable for use in the preparation of potassium trifluoro(2-methyl-1,3-dithiano)borate.

Solubility
Not miscible or difficult to mix in water. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6007-26-7
Molecular Formula C5H10S2
Molecular Weight (g/mol) 134.255
MDL Number MFCD00014652
InChI Key KXROTPXCYDXGSC-UHFFFAOYSA-N
Synonym 1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane
PubChem CID 22329
IUPAC Name 2-methyl-1,3-dithiane
SMILES CC1SCCCS1

Specifications

Density 1.212
Boiling Point 78°C to 80°C (9 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.5615
Quantity 5 g
Beilstein 102766
Solubility Information Not miscible or difficult to mix in water. Soluble in alcohol.
Formula Weight 134.25
Percent Purity 98%
Chemical Name or Material 2-Methyl-1,3-dithiane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.