missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dithiane, 97%, Thermo Scientific™

Product Code. 11309211 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
Packaging:
Glass bottle
Unit Size:
1mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11309211 5 mL Glass bottle 5mL
11936861 1 mL Glass bottle 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11309211 Supplier Thermo Scientific Acros Supplier No. 443260050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 6007-26-7
InChI Key KXROTPXCYDXGSC-UHFFFAOYSA-N
Synonym 1,3-dithiane, 2-methyl, m-dithiane, 2-methyl, 2-methyl-m-dithiane, methyl-2 dithiane-1,3, methyl-2 dithiane-1,3 french, acmc-20anrf, 2-methyl-1,3-dithian, 2-methyl-1,3 dithiane, 5-19-01-00053 beilstein handbook reference, 2-methyl-1,3-dithiane
PubChem CID 22329
IUPAC Name 2-methyl-1,3-dithiane
SMILES CC1SCCCS1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Brown
Density 1.1200g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Molecular Formula C5H10S2
Quantity 5 mL
Molecular Weight (g/mol) 134.255
Formula Weight 134.26
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 2-Methyl-1, 3-dithiane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.