missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-dioxolane, 98%

Product Code. p-7020761
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11495203 10 g 10g
11405213 50 g 50g
11415213 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11495203 Supplier Thermo Scientific Alfa Aesar Supplier No. A12824.09
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 497-26-7 | C4H8O2 | 88.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 497-26-7
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00003210
InChI Key HTWIZMNMTWYQRN-UHFFFAOYSA-N
Synonym 1,3-dioxolane, 2-methyl, 2-methyldioxole, 2-methyldioxolane, methyldioxolane, methyl dioxolane, 2-methyl-1,3-dioxacyclopentane, dioxolane, methyl, unii-94w0r6v90a, acetaldehyde ethylene acetal, methyl-1,3-dioxolane
PubChem CID 10342
IUPAC Name 2-methyl-1,3-dioxolane
SMILES CC1OCCO1

Specifications

Density 0.982
Boiling Point 80°C to 83°C
Flash Point −2°C (28°F)
Refractive Index 1.398
Quantity 10 g
UN Number UN1993
Beilstein 102520
Formula Weight 88.11
Percent Purity 98%
Chemical Name or Material 2-Methyl-1,3-dioxolane
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-1,3-dioxolane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-methyl-1,3-dioxolane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.