missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-cyclopentanedione, 98%

Product Code. 10081710 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10081710 25 g Glass bottle 25g
13428378 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10081710 Supplier Thermo Scientific Acros Supplier No. 152320250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 765-69-5 | C6H8O2 | 112.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 765-69-5
Molecular Formula C6H8O2
Molecular Weight (g/mol) 112.13
MDL Number MFCD00001406
InChI Key HXZILEQYFQYQCE-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclopentanedione, 1,3-cyclopentanedione, 2-methyl, 2-methyl-1,3-cyclopentadione, 2-methyl-cyclopentane-1,3-dione, unii-c9lg5vp01c, c9lg5vp01c, methylcycle-d, pubchem2206, acmc-209p4n, benzil-related compound, 46
PubChem CID 13005
IUPAC Name 2-methylcyclopentane-1,3-dione
SMILES CC1C(=O)CCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 212.0°C to 216.0°C
Color Beige to Tan
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol
Formula Weight 112.13
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methyl-1, 3-cyclopentanedione, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.