missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1,3-cyclohexanedione, 98+%

Product Code. 10418670 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
200 g
Packaging:
Glass bottle
Unit Size:
100g
200g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10418670 25 g Glass bottle 25g
10428670 100 g Glass bottle 100g
10138053 200 g Glass bottle 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10418670 Supplier Thermo Scientific Acros Supplier No. 126590250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1193-55-1 | C7H10O2 | 126.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1193-55-1
Molecular Formula C7H10O2
Molecular Weight (g/mol) 126.16
MDL Number MFCD00001587
InChI Key VSGJHHIAMHUZKF-UHFFFAOYSA-N
Synonym 2-methyl-1,3-cyclohexanedione, 1,3-cyclohexanedione, 2-methyl, 1,3-dimethylcycloadipic ketone, 2-methyl-1,3-cyclohexandione, 1-methyl-2,6-cyclohexanedione, ghl.pd_mitscher_leg0.98, 2-methylcyclohexan-1,3-dion, 2-methyl-cyclohexane-1,3-dione, pubchem15384, acmc-209a2g
PubChem CID 70945
IUPAC Name 2-methylcyclohexane-1,3-dione
SMILES CC1C(=O)CCCC1=O

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point >204°C
Color Brown to White
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3C6H7(=O)2
Quantity 25 g
Beilstein 07, 558
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol
Formula Weight 126.15
Percent Purity 98+%
Physical Form Crystalline Powder, Powder or Crystals
Chemical Name or Material 2-Methyl-1, 3-cyclohexanedione
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methyl-1,3-cyclohexanedione
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.