missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyresorcinol, TRC

Product Code. 30481042
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
500mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30481042 500 mg 500mg
30468233 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30481042 Supplier Toronto Research Chemicals Supplier No. TRCM267600500MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Methoxyresorcinol
CAS 29267-67-2
Molecular Formula C7 H8 O3
Quantity 500 mg
InChI Formula InChI=1S/C7H8O3/c1-10-7-5(8)3-2-4-6(7)9/h2-4,8-9H,1H3
SMILES COc1c(O)cccc1O
IUPAC Name 2-methoxybenzene-1,3-diol
Molecular Weight (g/mol) 140.14
Formula Weight 140.0473
Product Title
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.