missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine, 98%

Product Code. 10021440 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10021440 100 g Glass bottle 100g
10743393 5 g Glass bottle 5g
10773701 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10021440 Supplier Thermo Scientific Acros Supplier No. 126101000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1628-89-3 | C6H7NO | 109.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1628-89-3
Molecular Formula C6H7NO
Molecular Weight (g/mol) 109.13
MDL Number MFCD00006262
InChI Key IWTFOFMTUOBLHG-UHFFFAOYSA-N
Synonym pyridine, 2-methoxy, 2-methoxy pyridine, pyridine, methoxy, 2-methoxy-pyridine, unii-geb38j8108, methyl 2-pyridyl ether, methoxypyridine, 6-methoxypyridine, methyl pyridinyl ether, pubchem13201
PubChem CID 74201
IUPAC Name 2-methoxypyridine
SMILES COC1=CC=CC=N1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0400g/mL
Boiling Point 142°C
Flash Point 32°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)OCH3
Refractive Index 1.5010 to 1.503
Quantity 100 g
Beilstein 21, 44
Specific Gravity 1.04
Solubility Information Solubility in water: insoluble in water. Other solubilities: soluble in alcohol,acetone and ether
Formula Weight 109.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Methoxypyridine, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.