missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridine-5-boronic acid, 95%

Product Code. 11324526
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324526 5 g 5g
11334526 250 mg 250mg
11314526 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324526 Supplier Thermo Scientific Alfa Aesar Supplier No. L20087.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 163105-89-3 | C6H8BNO3 | 152.94 g/mol

2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxypyridine-5-boronic acid is used in cosmetics, coatings candles.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163105-89-3
Molecular Formula C6H8BNO3
Molecular Weight (g/mol) 152.94
MDL Number MFCD02093044
InChI Key DHADXDMPEUWEAS-UHFFFAOYSA-N
Synonym 2-methoxy-5-pyridineboronic acid, 6-methoxypyridin-3-yl boronic acid, 2-methoxypyridine-5-boronic acid, 2-methoxy-5-pyridinylboronic acid, 6-methoxypyridine-3-boronic acid, 6-methoxypyridin-3-yl-3-boronic acid, 6-methoxy-pyridine-3-boronic acid, 6-methoxy-3-pyridinylboronic acid, 6-methoxy-3-pyridylboronic acid, 6-methoxypyridin-3-yl boranediol
PubChem CID 2734368
IUPAC Name (6-methoxypyridin-3-yl)boronic acid
SMILES COC1=CC=C(C=N1)B(O)O

Specifications

Melting Point 132°C to 133°C
Quantity 5 g
Beilstein 9119950
Solubility Information Soluble in water.
Formula Weight 152.95
Percent Purity 95%
Chemical Name or Material 2-Methoxypyridine-5-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.