missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypyridin-3-amine, 97%, Thermo Scientific™

Product Code. 10623835
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10623835 25 g Amber Glass Bottle 25g
10416872 1 g Amber Glass Bottle 1g
10612083 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10623835 Supplier Thermo Scientific Maybridge Supplier No. SPB07839EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 20265-38-7
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.143
MDL Number MFCD00833386
InChI Key YXFAOWYMDGUFIQ-UHFFFAOYSA-N
Synonym 3-amino-2-methoxypyridine, 2-methoxy-3-aminopyridine, 2-methoxy-pyridin-3-ylamine, 3-pyridinamine, 2-methoxy, 2-methoxy-3-pyridylamine, 2-methoxy-3-pyridinamine, 2-methoxypyridine-3-amine, pubchem1269, 2-methoxy pyridin-3-amine
PubChem CID 2737488
IUPAC Name 2-methoxypyridin-3-amine
SMILES COC1=C(C=CC=N1)N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 25 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Methoxypyridin-3-amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.