missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Methoxypyrazine, 98%, Thermo Scientific™

Product Code. 11444043
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11444043 5 g 5g
11434043 1 g 1g
11454043 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11444043 Supplier Alfa Aesar™ Supplier No. A10297.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3149-28-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.116
MDL Number MFCD00006126
InChI Key WKSXRWSOSLGSTN-UHFFFAOYSA-N
Synonym pyrazine, methoxy, methoxypyrazine, 2-methoxy-1,4-diazine, pyrazine, 2-methoxy, 2-methoxy-pyrazine, unii-ryd35t7f4t, 2-methoxy pyrazine, fema no. 3302, ryd35t7f4t, methoxy-pyrazine
PubChem CID 18467
IUPAC Name 2-methoxypyrazine
SMILES COC1=NC=CN=C1

Specifications

Density 1.143
Boiling Point 147°C to 149°C
Flash Point 45°C (113°F)
Refractive Index 1.509
Quantity 5 g
UN Number UN1993
Beilstein 2241
Formula Weight 110.12
Percent Purity 98%
Chemical Name or Material 2-Methoxypyrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.