missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene, 97%, stabilized

Product Code. p-3736881 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10551241

Brand: Thermo Scientific Acros 174352500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 116-11-0 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.7800g/mL
Boiling Point 35°C to 36°C
Flash Point -29°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C(OCH3)=CH2
Refractive Index 1.3810 to 1.384
Quantity 250 mL
Beilstein 01, IV, 2075
Fieser 02,230; 05,360; 11,329; 12,317
Merck Index 15, 6073
Specific Gravity 0.78
Solubility Information Solubility in water: negligible
Stabilizer 0.5% max. potassium carbonate
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Methoxypropene, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methoxypropene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H224-Extremely flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P240-Ground/bond container and receiving equipment.
  • P243-Take precautionary measures against static discharge.
  • P264-Wash thoroughly after handling.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.