missing translation for 'onlineSavingsMsg'
Learn More

(2-Methoxyphenoxy)acetic acid, 98%, Thermo Scientific™

Product Code. 11905141 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11905141 10 g 10g
11915141 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11905141 Supplier Thermo Scientific Acros Supplier No. 442230100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 1878-85-9
Molecular Formula C9H10O4
Molecular Weight (g/mol) 182.18
InChI Key IHONYPFTXGQWAX-UHFFFAOYSA-N
Synonym 2-methoxyphenoxy acetic acid, 2-2-methoxyphenoxy acetic acid, 2-methoxyphenoxyacetic acid, guaiacoxyacetic acid, acetic acid, 2-methoxyphenoxy, o-methoxyphenoxy acetic acid, unii-7xvz1x365a, acide o-methoxyphenoxyacetique french, o-methoxy phenoxyacetic acid, acetic acid, o-methoxyphenoxy
PubChem CID 15882
IUPAC Name 2-(2-methoxyphenoxy)acetic acid
SMILES COC1=CC=CC=C1OCC(=O)O

Specifications

Melting Point 122°C to 123°C
Assay Percent Range 97.5% min. (HPLC)
Quantity 10 g
Formula Weight 182.18
Percent Purity 98%
Chemical Name or Material (2-Methoxyphenoxy)acetic acid
compliance-icons
Product Identifier
  • (2-Methoxyphenoxy)acetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.