missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyphenol, 98+%

Product Code. 11497787
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11497787 250 g 250g
11487787 1000 g 1000g
11407797 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11497787 Supplier Thermo Scientific Alfa Aesar Supplier No. A16319.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methoxyphenol, >-98%, C7H8O2, CAS Number-90-05-1, methylcatechol, guaiastil, 2-hydroxyanisole, 1-hydroxy-2-methoxybenzene, pyroguaiac acid, o-methoxyphenol, pyrocatechol monomethyl ether, phenol, 2-methoxy, guaicol, guaiacol, 1000g, >98%, 508112, 204 deg.C to 206 deg.C, CHEBI:28591, 1.129 | CAS: 90-05-1 | C7H8O2 | 124.139 g/mol

2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methoxyphenol acts as an expectorant. It is used as an anti-oxidant as well as an anti-skinning agent for paints. It serves as a precursor to flavorants such as eugenol and vanillin. Further, it is used as an intermediate in the chemical synthesis of active pharmaceuticals, flavoring and perfumery products. In addition to this, it is also used as an indicator in chemical reactions that produce oxygen.

Solubility
Miscible with carbon tetrachloride, alcohol, chloroform, ether, oils, glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.

Notes
Air and light sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-05-1
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.139
MDL Number MFCD00002185
InChI Key LHGVFZTZFXWLCP-UHFFFAOYSA-N
Synonym guaiacol, o-methoxyphenol, 2-hydroxyanisole, phenol, 2-methoxy, pyrocatechol monomethyl ether, methylcatechol, guaiastil, guaicol, 1-hydroxy-2-methoxybenzene, pyroguaiac acid
PubChem CID 460
ChEBI CHEBI:28591
IUPAC Name 2-methoxyphenol
SMILES COC1=CC=CC=C1O

Specifications

Melting Point 28°C to 31°C
Density 1.129 g/mL
Boiling Point 204°C to 206°C
Flash Point 82°C (179°F)
Assay Percent Range >98%
Refractive Index 1.543
Quantity 250 g
Beilstein 508112
Sensitivity Air and light sensitive
Merck Index 14,4553
Solubility Information Miscible with carbon tetrachloride,alcohol,chloroform,ether,oils,glacial acetic acid and sodium hydroxide solution. Slightly miscible with water and petroleum ether.
Formula Weight 124.14
Percent Purity ≥98%
Chemical Name or Material 2-Methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.